20.90 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Certifications/Compliance | CE, ISO 11193-1, EN420, 89/686/EEC, EN374-2: 2003, EN374-3: 2003, MDD 93/42/EEC, Class 1, EN455-1, EN455-2 and EN455-3, ASTM F1671, ISO 21171 |
---|---|
AQL | 0.65 |
Color | Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | 7.5/8 |
Sterility | Non-sterile |
Thickness (Metric) | 0.07 mm |
Finger Style | Textured |
Glove Cuff Style | Beaded/Rolled |
Model | 93-833 |
Packaging Type | Dispenser box |
Quantity | 250/Pk. |
Tensile Strength | 14 to 33 MPa |
Length (Metric) | 24 cm |
Width (Metric) | 96 mm |