472.60 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Use promo code "24107" to get your promotional price.
Classification | Animal Origin-Free |
---|---|
Content And Storage | Contains one vial of Lipofectamine™ 3000 Reagent and one vial of P3000TM Enhancer Reagent. Store at 2–8°C. Do not freeze. |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Transfection Technique | Lipid-based Transfection |
For Use With (Application) | Transfection |
Green Features | Sustainable packaging |
Product Line | Lipofectamine™ |
Product Type | Transfection Reagent |
Quantity | 0.75 mL |
Serum Compatible | Yes |