Thermo Scientific™ 96-well/Flat Bottom Block
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Overview
80.61 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.40000kg
Product Code. 15312235
Quantity Price
1 £ 92.55 / Each
Estimated Shipment
18-06-2025
Add to basket
Description and Specification
Specification
Description 96-well/flat bottom plate block
Capacity (Metric) 0.2 mL
For Use With (Equipment) Thermo Scientific™ Compact Dry Bath/Block Heater
Lighting LED
No. of Places 96
Type Block
Dimensions (L x W x H) 108 x 72 x 18 mm (4.3 x 2.8 x 0.7 in.)
Depth (Metric) 13 mm
Warranty: 2 years
Special Offers