Thermo Scientific™ TurboFect™ Transfection Reagent
Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.
Overview
231.44 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15325016
Quantity Price
1 £ 270.0 / 1mL
In stock, ready to ship: 6
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Type Transfection Reagent
Quantity 1 mL
Classification Chemically Defined
Compatible Cells Primary Cell, Endothelial Cell, Epithelial Cell, Myeloma Cell, Cell Lines
Format 6-Well Plate, 12-Well Plate, 24-Well Plate, 48-Well Plate, 96-Well Plate, Flasks
Transfection Technique Chemical Transfection
Product Line TurboFect
For Use With (Application) Transfection
The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery in vivo. Reagent causes no detectable inflamatory response and is suitable for DNA administration via various routes, including intravenous, intraperitoneal, intratumoral injection, etc.
  • No detectable inflamatory response
  • Gene delivery via multiple routes of administration
  • Reproducible results
  • Functionally tested in mice and rats
Recommended for:
DNA transfection
siRNA-DNA co-transfection