15.64 GBP valid until 2025-12-31
Use promo code "24341" to get your promotional price.
Use promo code "24341" to get your promotional price.
Additive | No Sodium Pyruvate |
---|---|
Classification | Animal Origin-Free |
Color | Clear |
Content And Storage |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
Packaging Type | Bottle |
Dilution | 1 X |
Product Line | Gibco™ |
Product Type | DPBS |
Quantity | 500 mL |
Solution Type | Dulbecco's Phosphate Buffered Saline |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 270 - 300 mOsm/kg |
pH | 7.0 to 7.3 |
Phenol Red Indicator | No Phenol Red |
Shipping Condition | Room Temperature |
Sterility | Sterile-filtered |
Form | Liquid |
Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |