Code:
NEW
Corning™ cellgro™ MEM Alpha
Additional Details:
Weight:
3.76000kg
Specification
Concentration | 1 X |
---|---|
Product Type | Minimum Essential Medium (MEM) alpha |
For Use With (Application) | To assess growth promotion and cytotoxicity characteristics |
Quantity | 6/Pk. |
Endotoxin Level | <0.25EU/mL |
Formulation | Phenol Red, Sodium Pyruvate, Sodium Bicarbonate |
Content And Storage | 2 to 8°C |