Code:
NEW
Thermo Scientific™ Block, 6 x 2.0 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Additional Details:
Weight:
0.11160kg
Specification
Description | 6 x 2.0mL block |
---|---|
Capacity (Metric) | 2 mL |
For Use With (Equipment) | Thermo Scientific™ Compact Dry Bath/Block Heater |
Lighting | LED |
No. of Places | 6 |
Type | Block |
Dimensions (L x W x H) | 36 x 54 x 30 mm (1.4 x 2.1 x 1.2 in.) |