Thermo Scientific™ Block, 6 x 15 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Overview
63.69 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.47520kg
Product Code. 15392225
Quantity Price
1 £ 72.72 / Each
Estimated Shipment
10-06-2025
Add to basket
Description and Specification
Specification
Description 6 x 15mL block
Capacity (Metric) 15 mL
For Use With (Equipment) Thermo Scientific™ Compact Dry Bath/Block Heater
Lighting LED
No. of Places 6
Type Block
Dimensions (L x W x H) 36 x 108 x 72 mm (1.4 x 4.3 x 2.8 in.)
Warranty: 2 years