Thermo Scientific Alfa Aesar 4-Nitrobenzenediazonium tetrafluoroborate, 97%
CAS: 456-27-9 | C6H4BF4N3O2 | 236.92 g/mol
Overview
22.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 456-27-9 Weight: 0.00100kg Transport: UN number : 2923 Chem class : 8 Pack group : II
Quantity 1 g
Disclaimers: RUO – Research Use Only
Product Code. 15431538
Quantity Price
1 £ 22.75 / 1g
In stock, ready to ship: 2
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 456-27-9
Molecular Formula C6H4BF4N3O2
Molecular Weight (g/mol) 236.92
MDL Number MFCD00012005
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
PubChem CID 68006
IUPAC Name 4-nitrobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
Specification
Chemical Name or Material 4-Nitrobenzenediazonium tetrafluoroborate
CAS 456-27-9
Melting Point 144°C to 148°C
Molecular Formula C6H4BF4N3O2
MDL Number MFCD00012005
Quantity 1 g
UN Number UN2923
Beilstein 3599869
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Synonym 4-nitrobenzenediazonium tetrafluoroborate,ccris 3641,unii-2uxo615r4o,p-nitrobenzenediazonium tetrafluoroborate,benzenediazonium, 4-nitro-, tetrafluoroborate 1-,p-nitrophenyldiazonium fluoborate,p-nitrobenzenediazonium fluoborate,4-nitrobenzenediazonium fluoborate,p-nitrobenzenediazonium fluoroborate,4-nitrobenzenediazonium fluoroborate
Solubility Information Soluble in acetonitrile and dimethyl sulfoxide.
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
IUPAC Name 4-nitrobenzenediazonium;tetrafluoroborate
Molecular Weight (g/mol) 236.92
PubChem CID 68006
Formula Weight 236.92
Percent Purity 97%
Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.
Solubility
Soluble in acetonitrile and dimethyl sulfoxide.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.