Micronova™ Stainless Steel Wire Brush
Durable stainless steel wire brushes with nylon bristles are available in a variety of sizes for cleaning parts, tubing and hard to reach areas.
Additional Details:
Weight:
1.00000kg
Specification
Product Type | J-Shaped Brush |
---|---|
Bristle Material | Nylon |
Bristle Type | Medium Stiffness |
Color-coded | Yes |
Handle Material | Stainless Steel Wire with Plastic Grip |
Handle Type | Acme Thread |
Tip Type | Tuft End |
Sterility | Non-sterile |
Appearance | Wire brush^ Flexbrush, flexible inchjinch shaped, stainless steel wire brush on plastic handle with acme connection. |
Autoclavable | Autoclavable |
Care Instructions | Item can be cleaned after use and hung up for optimal drying and sanitation purposes. Flexibility of the head allows users to 'shape' the head for custom cleaning applications |
Certifications/Compliance | ISO 9001 Certified Facility; Certificate of Conformance; FDA Approved Materials |
Color | White |
Content And Storage | Store in a dry area, at ambient temperature with ideal air flow. |
Diameter (English) Bristle | 2.25 in. |
Diameter (Metric) Bristle | 5.72 cm |
Dimensions (L x W x H) | 16 x 2.2 x 2.2 in. (40.6 x 5.7 x 5.7 cm) |
Disposable | Reusable |
Filtered | Non-filtered |
Flammable | No |
For Use With (Application) | Brushes flex to clean ducting, piping exteriors or vessel interiors. |
For Use With (Equipment) | STAH or STAEH series handles; FlexBrushTM Covers |
Includes | Without Handle |
Length (English) | 16 in. |
Length (English) Brush | 12 in. |
Length (English) Handle | 4 in. |
Length (Metric) | 40.6 cm |
Length (Metric) Brush | 30.4 cm |
Length (Metric) Handle | 10.2 cm |
Material | Nylon, Stainless Steel (304), Plastic |
Packaging | Each brush is individually packaged |
Quantity | 12/Cs. |
Resistant To | Standard disinfectants and cleaning solutions |
Size | 16 x 10 x 8 in. |
Weight (English) | 9 lb. |
Weight (Metric) | 4.08 kg |
Width (English) Bristle | 2.25 in. |
Width (Metric) Bristle | 5.7 cm |