Code:
NEW
Honeywell Riedel-de Haën Tert-Butanol, puriss. p.a. ACS Reagent, ≥99.7% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS reagent,≥99.7% (GC)
Additional Details:
CAS Number:
75-65-0
Weight:
1.97500kg
Transport:
UN number : 1120
Chem class : 3
Pack group : II
Quantity | 2.5 L |
---|---|
Packaging | Poly Bottle |
Chemical Identifiers
CAS | 75-65-0 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00004464 |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
PubChem CID | 6386 |
ChEBI | CHEBI:45895 |
IUPAC Name | 2-methylpropan-2-ol |
SMILES | CC(C)(C)O |
Specification
Chemical Name or Material | Tert-Butanol |
---|---|
Melting Point | 25°C |
CAS | 75-65-0 |
Color | Colorless |
Density | 0.775g/mL at 25°C |
Boiling Point | 83°C |
Quantity | 2.5 L |
Explosion Limit | 0.08 |
Flash Point | 11°C (51.8°F) |
Ignition Point | 896°F |
Packaging | Poly Bottle |
Physical Form | Liquid |
Refractive Index | n20/D 1.387(lit.) |
Molecular Formula | C4H10O |
Linear Formula | (CH3)3COH |
MDL Number | MFCD00004464 |
Vapor Pressure | 44mmHg (26°C), 31mmHg (20°C) |
UN Number | UN1120 |
Beilstein | 906698 |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
SMILES | CC(C)(C)O |
IUPAC Name | 2-methylpropan-2-ol |
Molecular Weight (g/mol) | 74.123 |
PubChem CID | 6386 |
ChEBI | CHEBI:45895 |
Formula Weight | 74.12g/mol |
Percent Purity | ≥99.7% (GC) |
Grade | ACS/puriss. p.a. |