Code:
NEW
Honeywell Research Chemicals Toluene, Laboratory Reagent, ≥99.3%, Honeywell™
Laboratory Reagent,≥99.3%
Additional Details:
CAS Number:
108-88-3
Weight:
5.19000kg
Transport:
UN number : 1294
Chem class : 3
Pack group : II
Quantity | 6 x 1 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Specification
Chemical Name or Material | Toluene |
---|---|
Melting Point | -95°C |
CAS | 108-88-3 |
Color | Colorless |
Density | 0.865g/mL at 25°C |
Boiling Point | 110.6°C |
Quantity | 6 x 1 L |
Explosion Limit | 0.07 |
Flash Point | 4°C (39.2°F) |
Ignition Point | 997°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | n/D 1.496(lit.) |
Molecular Formula | C7H8 |
Linear Formula | C6H5CH3 |
MDL Number | MFCD00008512 |
Vapor Pressure | 22mmHg (20°C), 26mmHg (25°C) |
UN Number | UN1294 |
Beilstein | 635760 |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
Synonym | methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
SMILES | CC1=CC=CC=C1 |
IUPAC Name | toluene |
Molecular Weight (g/mol) | 92.14 |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
Formula Weight | 92.14g/mol |
Percent Purity | ≥99.3% |
Grade | Laboratory Reagent |