BINDER™ Series ED Avantgarde.Line - Drying and Heating Chamber, with Natural Convection
The strengths of an ED Series Drying Chamber include routine drying and sterilization tasks up to 300°C. Thanks to the natural convection, all thermal processes which use this drying chamber are highly efficient. The ED series ensures fast, uniform drying.
Overview
887.92 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Additional Details:
Additional Details: Weight: 50.00000kg
Product Code. 15622126
Quantity Price
1 £ 1452.3 / Each
In stock, ready to ship: 147
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
  • Warranty
  • Special Offers
Description and Specification
Specification
Capacity (English) 2 cu. ft.
Chamber Material Stainless Steel
Controller Type Controller with LCD display
Convection Type Natural
Description ED056-230 V, Standard, Series ED Avantgarde.Line - Drying and heating chambers with natural convection, 57 L, 230 V 1 ∼ 50/60 Hz
Capacity (Metric) 57 L
Temperature (Metric) Ambient +5° to 300°C
Type Drying and Heating Chamber
Display LCD
Material Stainless Steel
Max. Shelf Load 30 kg (66.1 lb.)
Model ED056-230 V
No. of Shelves 2 std./4 max.
Plug Type NEMA 5-20P
Dimensions (D x W x H) Interior 360 x 380 x 420 mm
Dimensions (L x W x H) 635 x 665 x 625 mm
Weight (Metric) 42 kg
Electrical Requirements 230 V, 1 Phase, 50/60 Hz, 1.05 kW, 6.3 A
Voltage 230 V
Wattage 1.05 kW
Amperage 6.3 A
Frequency 50/60 Hz
Convection Technology APT.Line Technology
No. of Doors 1
  • Temperature range: +5°C above ambient temperature to +300°C
  • Up to 30% lower energy consumption compared to conventional units on the market
  • High temperature accuracy thanks to APT.line™ technology
  • Natural convection
  • Controller with LCD display
  • Electromechanical control of the exhaust air flap
  • 2 chrome-plated racks
  • Class 2 integrated independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Ergonomic handle design
  • USB port for recording data
Alternative Products
Accessories and Spare Parts
Warranty
Special Offers