BINDER™ Series FD Avantgarde.Line - Drying and Heating Chamber, with Forced Convection
An FD Series Avantgarde.Line heating oven is used when fast drying and sterilization is required. Thanks to its fully homogeneous temperature distribution, quick dynamics and powerful fan, this heating oven saves valuable time.
Overview
Additional Details:
Additional Details: Weight: 66.00000kg
Product Code. 15632116
Quantity Price
1 £ 2570.0 / Each
In stock, ready to ship: 281
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
  • Warranty
  • Special Offers
Description and Specification
Specification
Capacity (English) 4.1 cu. ft.
Chamber Material Stainless Steel
Controller Type Controller with LCD display
Convection Type Forced
Description FD115-230 V, Standard, Series FD Avantgarde.Line - Drying and heating chambers with forced convection, 116 L, 230 V 1 ∼ 50/60 Hz
Capacity (Metric) 116 L
Temperature (Metric) Ambient +10° to 300°C
Type Drying and Heating Chamber
Display LCD
Material Stainless Steel
Max. Shelf Load 30 kg (66.1 lb.)
Model FD115-230 V
No. of Shelves 2 std./5 max.
Plug Type NEMA 5-20P
Dimensions (D x W x H) Interior 550 x 385 x 550 mm
Dimensions (L x W x H) 835 x 760 x 705 mm
Weight (Metric) 57 kg
Electrical Requirements 230 V, 1 Phase, 50/60 Hz, 1.3 kW, 6.3 A
Voltage 230 V
Wattage 1.3 kW
Amperage 6.3 A
Frequency 50/60 Hz
Convection Technology APT.Line Technology
No. of Doors 1
  • Up to 30% lower energy consumption compared to conventional units on the market
  • High temperature accuracy thanks to APT.line™ technology
  • Forced convection
  • Controller with LCD display
  • Electromechanical control of the exhaust air flap
  • 2 chrome-plated racks
  • Class 2 integrated independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Ergonomic handle design
  • USB port for recording data
Alternative Products
Accessories and Spare Parts
Warranty
Special Offers