Honeywell Fluka Sodium carbonate, Powder, ≥99.5%, ACS reagent, Honeywell Fluka™
Quantity | 2.5 kg |
---|---|
Packaging | Poly Bottle |
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Specification
Chemical Name or Material | Sodium carbonate |
---|---|
Melting Point | 851°C (literature) |
Name Note | Powder, ≥99.5%, ACS reagent |
CAS | 497-19-8 |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Physical Form | Powder |
Quantity | 2.5 kg |
Assay Percent Notes | ex ignited substance |
Molecular Formula | CNa2O3 |
Linear Formula | Na2CO3 |
MDL Number | MFCD00003494 |
Beilstein | 4154566 |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
Synonym | sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
IUPAC Name | disodium carbonate |
Molecular Weight (g/mol) | 105.99 |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
Formula Weight | 105.99 |
Percent Purity | >99.5% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Density | 2.53 g/cm3 |
Technical documents
[Sodium carbonate]
[Warning]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=52, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=53, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : sodium carbonate}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- Sodium carbonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319- Causes serious eye irritation.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : sodium carbonate