Honeywell Research Chemicals Ethylene glycol, Reagent Grade, ≥99%, Honeywell™
Quantity | 2.5 L |
---|---|
Packaging | Poly Bottle |
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
MDL Number | MFCD00002885 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Specification
Chemical Name or Material | Ethylene glycol |
---|---|
Melting Point | -13°C (literature) |
Name Note | Reagent Grade, ≥99% |
CAS | 107-21-1 |
CAS Max % | 100.0000% |
Density | 1.113 g/mL at 25 °C(lit.) |
Boiling Point | 197°C |
Quantity | 2.5 L |
Explosion Limit | 0.153 |
Flash Point | 111°C |
Ignition Point | 752°F |
Packaging | Poly Bottle |
Molecular Formula | C2H6O2 |
Linear Formula | HOCH2CH2OH |
MDL Number | MFCD00002885 |
Vapor Pressure | 0.08mmHg (20°C) |
UN Number | UN3082 |
Beilstein | 505945 |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
Solubility Information | Miscible in water |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
SMILES | C(CO)O |
IUPAC Name | ethane-1,2-diol |
Molecular Weight (g/mol) | 62.068 |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
Formula Weight | 62.07 |
Percent Purity | >99% |
Grade | Reagent |
Technical documents
[Ethylene glycol]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=STOT RE 2, sequence=2, description=Specific target organ toxicity after repeated exposure Category 2}]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H373, sequence=2, description=May cause damage to organs.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P308+P313, sequence=54, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethylene glycol}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Ethylene glycol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302- Harmful if swallowed.
- H373- May cause damage to organs.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : Ethylene glycol