Honeywell Riedel-de Haën tert-Butyl methyl ether, For pesticide residue analysis, Honeywell Riedel-de Haën™
Quantity | 1 L |
---|
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Specification
Chemical Name or Material | tert-Butyl methyl ether |
---|---|
CAS | 1634-04-4 |
Density | 0.74g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.151 |
Flash Point | −33°C (−27.4°F) |
Ignition Point | 705°F |
Molecular Formula | C5H12O |
Linear Formula | (CH3)3COCH3 |
MDL Number | MFCD00008812 |
UN Number | UN2398 |
Beilstein | 1730942 |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
SMILES | CC(C)(C)OC |
IUPAC Name | 2-methoxy-2-methylpropane |
Molecular Weight (g/mol) | 88.15 |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
Formula Weight | 88.15g/mol |
Percent Purity | ≥99.7% |
Grade | CHROMASOLV™ |
Technical documents
[tert-Butyl methyl ether]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=SKIN IRRIT. 2, sequence=2, description=Skin corrosion/irritation Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H315, sequence=2, description=Causes skin irritation.}]
[{code=P243, sequence=51, description=Take precautionary measures against static discharge.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P308+P313, sequence=54, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : tert-butyl methyl ether}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- tert-Butyl methyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H315- Causes skin irritation.
Precautionary Statement
- P243- Take precautionary measures against static discharge.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : tert-butyl methyl ether