Use promo code "24334" to get your promotional price.
Honeywell Riedel-de Haën Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™
Use promo code "24334" to get your promotional price.
Quantity | 6 x 1 L |
---|
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Specification
Chemical Name or Material | Acetonitrile |
---|---|
Melting Point | -48°C |
Name Note | for HPLC |
Absorbance | HPLC-gradient/254nm ≤0.5mAU |
CAS | 75-05-8 |
Density | 0.786g/mL at 25°C |
Boiling Point | 81°C to 82°C |
Quantity | 6 x 1 L |
Refractive Index | n20/D 1.344 |
Molecular Formula | C2H3N |
Linear Formula | CH3CN |
MDL Number | MFCD00001878 |
Vapor Pressure | 72.8mmHg (20°C) |
Beilstein | 741857 |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Molecular Weight (g/mol) | 41.053 |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
Formula Weight | 41.05g/mol |
Percent Purity | ≥99.9% |
Grade | CHROMASOLV™ Gradient |
Technical documents
[Acetonitrile]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H312, sequence=3, description=Harmful in contact with skin.}, {code=H319, sequence=4, description=Causes serious eye irritation.}, {code=H332, sequence=5, description=Harmful if inhaled.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=52, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=54, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=56, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=57, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : acetonitrile}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : acetonitrile