Honeywell Fluka Sodium hypochlorite solution, purum, ≈10% (RT), Honeywell Fluka™
Quantity | 2.5 L |
---|
Chemical Identifiers
CAS | 7681-52-9 |
---|---|
Molecular Formula | ClNaO |
Molecular Weight (g/mol) | 74.439 |
MDL Number | MFCD00011120 |
InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
PubChem CID | 23665760 |
ChEBI | CHEBI:32146 |
IUPAC Name | sodium;hypochlorite |
SMILES | [O-]Cl.[Na+] |
Specification
Chemical Name or Material | Sodium hypochlorite solution |
---|---|
CAS | 7681-52-9 |
Quantity | 2.5 L |
Molecular Formula | ClNaO |
Linear Formula | NaClO |
MDL Number | MFCD00011120 |
UN Number | UN1791 |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
SMILES | [O-]Cl.[Na+] |
IUPAC Name | sodium;hypochlorite |
Molecular Weight (g/mol) | 74.439 |
PubChem CID | 23665760 |
ChEBI | CHEBI:32146 |
Formula Weight | 74.44g/mol |
Percent Purity | 10% (RT) |
Grade | purum |
Concentration or Composition (by Analyte or Components) | ≈10% (RT) |
Density | 1.21-1.26g/mL at 20°C |
Technical documents
[Sodium hypochlorite solution]
[Danger]
[{code=MET. CORR. 1, sequence=1, description=Substances and mixtures corrosive to metals Category 1}, {code=SKIN CORR. 1B, sequence=2, description=Skin corrosion/irritation Category 1 B}, {code=EYE DAM. 1, sequence=3, description=Serious eye damage/eye irritation Category 1}, {code=AQUATIC ACUTE 1, sequence=4, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 2, sequence=5, description=LONG-TERM AQUATIC HAZARD Chronic 2}]
[{code=H290, sequence=1, description=May be corrosive to metals.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H410, sequence=3, description=Very toxic to aquatic life with long lasting effects.}, {code=EUH031, sequence=91, description=Contact with acids liberates toxic gas.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=53, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P301+P330+P331, sequence=54, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=55, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=56, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=57, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=58, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : sodium hypochlorite,solution}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS09, sequence=2, description=ENVIRONMENT}]
Product Identifier
- Sodium hypochlorite solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H290- May be corrosive to metals.
- H314- Causes severe skin burns and eye damage.
- H410- Very toxic to aquatic life with long lasting effects.
- EUH031- Contact with acids liberates toxic gas.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : sodium hypochlorite,solution