BINDER™ Series FD Classic.Line - Drying and Heating Chamber, with Forced Convection
A BINDER FD series heating oven is always used when fast drying and sterilization is required. Thanks to its fully homogeneous temperature distribution, quick dynamics and powerful fan, this heating oven saves valuable time.
Overview
1373.93 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Additional Details:
Additional Details: Weight: 32.00000kg
Product Code. 15682086
Quantity Price
1 £ 2245.0 / Each
In stock, ready to ship: 41
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
  • Warranty
  • Special Offers
Description and Specification
Specification
Certifications/Compliance ASTM D5374, DIN 12880
Description FD023-230 V, Standard, Series FD Classic.Line - Drying and heating chambers with forced convection, 20 L, 230 V 1 ∼ 50/60 Hz
Capacity (Metric) 20 L
Temperature (Metric) 5°C to 300°C
Type Drying and Heating Chamber
Display LED
Model FD023-230 V
No. of Shelves 2 (Standard), 4 (Maximum)
Ports 10/30/50/100 mm
Safety Features User friendly and easy to operate all-digital PID controller technology
Temperature Uniformity 150°C
Timer 0 to 99 hr
Uniformity 2.2°C
Dimensions (D x W x H) Interior 222 x 300 x 330 mm
Dimensions (L x W x H) 435 x 605 x 495 mm
Width (Metric) 435 mm
Height (Metric) 495 mm
Weight (Metric) 27 kg
Voltage 230 V
Wattage 0.8 kW
Amperage 10 A
Frequency 50/60 Hz
  • Temperature range: +5°C above ambient temperature to +300 °C
  • APT.line™ preheating chamber technology
  • Forced convection
  • Adjustable exhaust air flap
  • Controller with timer functions
  • 2 chrome-plated racks
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
Alternative Products
Accessories and Spare Parts
Warranty
Special Offers