HUBER KISS 212B Heating Bath Circulator
Heating bath circulators with a stainless steel bath allow the temperature control of internal and external thermoregulation tasks. Items can be directly thermoregulated by placing them in a thermostat bath and the control of externally connected applications is also possible.
Additional Details:
Weight:
11.00000kg
Specification
Controller Type | KISS |
---|---|
Capacity (Metric) | 12 L |
Certifications/Compliance | III/FL (DIN 12876) |
Pump Type | M16x1 AG Male |
Temperature Range | 25°C to 200°C |
Type | KISS 212 B Heating Bath Thermostat |
Heating Capacity | 1.5 - 2.1 kW |
Material | Stainless Steel |
Max. Pressure | 14 L/min; 0.25 bar |
Model | KISS 212 B |
Temperature Stability | ±0.05°C |
Depth (Metric) | 375 mm |
Weight (English) | 24.2 lb. |
Weight (Metric) | 11 kg |
Voltage | 208/240 V |
Amperage | 10 A |
Frequency | 50/60 Hz |