Honeywell Research Chemicals Chloroform, ≥99.5%, Honeywell™
Quantity | 500 mL |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Specification
Chemical Name or Material | Chloroform |
---|---|
CAS | 67-66-3 |
Melting Point | -61.3°C |
Color | Colorless |
Density | 1.492g/mL at 25°C |
Boiling Point | 61°C (1013 hPa) |
Odor | Sweet |
Packaging | Glass bottle |
Molecular Formula | CHCl3 |
MDL Number | MFCD00000826 |
Quantity | 500 mL |
UN Number | UN1888 |
Beilstein | 1731042 |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
Synonym | trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
SMILES | ClC(Cl)Cl |
IUPAC Name | chloroform |
Stabilizer | Amylenes |
Molecular Weight (g/mol) | 119.37 |
PubChem CID | 6212 |
Vapor Density | 4.1 (vs air) |
ChEBI | CHEBI:35255 |
Formula Weight | 119.38g/mol |
Vapor Pressure | 160mmHg (20°C) |
Percent Purity | ≥99.5% |
Physical Form | Liquid |
Technical documents
[Chloroform]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=ACUTE TOX. 3, sequence=2, description=Acute toxicity Category 3}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 2, sequence=4, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=5, description=Specific target organ toxicity Category 3}, {code=STOT RE 1, sequence=6, description=Specific target organ toxicity Category 1}, {code=CARC. 2, sequence=7, description=Carcinogenicity Category 2}, {code=REPR. 2, sequence=8, description=Reproductive toxicity Category 2}]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H331, sequence=4, description=Toxic if inhaled.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}, {code=H351, sequence=6, description=Suspected of causing cancer.}, {code=H361d, sequence=7, description=Suspected of damaging the unborn child.}, {code=H372, sequence=8, description=Causes damage to organs.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=53, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P301+P330+P331, sequence=54, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=55, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=56, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=57, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=58, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=GHS06, sequence=1, description=SKULL & CROSSBONES}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Chloroform
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H331- Toxic if inhaled.
- H336- May cause drowsiness or dizziness.
- H351- Suspected of causing cancer.
- H361d- Suspected of damaging the unborn child.
- H372- Causes damage to organs.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.