Use promo code "24127" to get your promotional price.
Honeywell Riedel-de Haën Cyclohexane, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Use promo code "24127" to get your promotional price.
Quantity | 1 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Specification
Chemical Name or Material | Cyclohexane |
---|---|
Melting Point | 6.4°C |
CAS | 110-82-7 |
Density | 0.779g/mL at 25°C |
Boiling Point | 81°C |
Quantity | 1 L |
Explosion Limit | 0.09 |
Flash Point | −18°C (−.4°F) |
Ignition Point | 500°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | n20/D 1.426(lit.) |
Molecular Formula | C6H12 |
MDL Number | MFCD00003814 |
Vapor Pressure | 168.8mmHg (37.7°C), 77mmHg (20°C) |
UN Number | UN1145 |
Beilstein | 1900225 |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
Synonym | hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
SMILES | C1CCCCC1 |
IUPAC Name | cyclohexane |
Molecular Weight (g/mol) | 84.16 |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
Formula Weight | 84.16g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Technical documents
[Cyclohexane]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ASP. TOX. 1, sequence=2, description=Aspiration toxicity Category 1}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}, {code=AQUATIC ACUTE 1, sequence=5, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 1, sequence=6, description=LONG-TERM AQUATIC HAZARD Chronic 1}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H304, sequence=2, description=May be fatal if swallowed and enters airways.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H336, sequence=4, description=May cause drowsiness or dizziness.}, {code=H410, sequence=5, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P260, sequence=52, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=54, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P301+P330+P331, sequence=55, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=56, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=57, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P308+P313, sequence=58, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H304- May be fatal if swallowed and enters airways.
- H315- Causes skin irritation.
- H336- May cause drowsiness or dizziness.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
-
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.
Buy Now