BINDER™ Series BD Avantgarde.Line - Standard-Incubator, with Natural Convection
The BD Avantgarde.Line Series is specially designed for gentle incubation of organisms, as well as conditioning of heat-sensitive media. The new design of the units and the homogeneous temperature distribution ensure the very best results.
Overview
1095.15 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Additional Details:
Additional Details: Weight: 48.00000kg
Product Code. 15692116
Quantity Price
1 £ 1855.0 / Each
In stock, ready to ship: 81
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
  • Warranty
  • Special Offers
Description and Specification
Specification
Capacity (English) 2 cu. ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Description BD056-230 V, Standard, Series BD Avantgarde.Line - Standard-Incubators with natural convection, 57 L, 230 V 1 ∼ 50/60 Hz
Autoclavable Not Autoclavable
Capacity (Metric) 57 L
Chamber Material Stainless Steel
Convection Technology APT.Line Technology
Convection Type Natural
Holds None
Lighting No Lighting
Material Stainless Steel
No. of Places 1
No. of Shelves 2
Plug Type 2-pin, CEE7/7. (If UK or Ireland, includes 3-pin UK plug.)
Temperature Range (Metric) Ambient +5° to 100°C
Type Standard Incubator
Display LCD
Door Style Glass Door
Model BD056-230 V
Dimensions (D x W x H) Interior 360 x 380 x 420 mm
Dimensions (L x W x H) 565 x 640 x 625 mm
Weight (Metric) 40 kg
Voltage 230 V
Wattage 0.3 kW
Amperage 6.3 A
Frequency 50/60 Hz
  • Temperature range: +5°C above ambient temperature to +100°C
  • High temperature accuracy thanks to APT.line™ technology
  • Natural convection
  • Controller with LCD display
  • Electromechanical control of the exhaust air flap
  • Inner door made of tempered safety glass
  • 2 chrome-plated racks
  • Class 3.1 integrated independent temperature safety device (DIN 12880) with visual alarm
  • Ergonomic handle design
  • USB port for recording data
Alternative Products
Accessories and Spare Parts
Warranty
Special Offers