Honeywell Fluka Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
Quantity | 4 x 2.5 L |
---|---|
Packaging | Plastic Bottle |
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Specification
Chemical Name or Material | Acetic acid |
---|---|
Name Note | Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% |
CAS | 64-19-7 |
Melting Point | 16.2°C (literature) |
Boiling Point | 118°C |
CAS Max % | 100.0000% |
Molecular Formula | C2H4O2 |
Linear Formula | CH3CO2H |
MDL Number | MFCD00036152 |
UN Number | UN2789 |
Beilstein | 506007 |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
Formula Weight | 60.05 |
Percent Purity | >99.8% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Density | 1.049 g/mL at 25 °C(lit.) |
Quantity | 4 x 2.5 L |
Flash Point | 40°C |
Packaging | Plastic Bottle |
Technical documents
[Acetic acid]
[Danger]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=SKIN CORR. 1A, sequence=2, description=Skin corrosion/irritation Category 1 A}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P284, sequence=52, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=56, description=IF exposed or concerned: Get medical advice/attention.}, {code=P280, sequence=57, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=58, description=IF ON SKIN: Wash with plenty of water/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Acetic acid
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226- Flammable liquid and vapour.
- H314- Causes severe skin burns and eye damage.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
-
Science Is Hard Enough Your Bioreagents Shouldn’t Be
Reliable performance. Special prices.Happier researchers.
Buy Now