285.30 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Autoclavable | Autoclavable |
---|---|
Color | Light Gray |
For Use With | Pipette Tip 20 μL |
Number of Channels | 1 |
Product Line | Reference 2 |
Type | Pipette |
Volume (Metric) | 2 to 20 μL |
For Use With (Equipment) | 20 μL pipette tips |
Includes | epT.I.P.S.™ Box 2.0 with 96 Tips |
Increments | 0.02 μL |
Color Code | Light Gray |