903.35 GBP valid until 2025-06-30
Use promo code "24072" to get your promotional price.
Use promo code "24072" to get your promotional price.
Capacity | 12 x 2 mL |
---|---|
Max. Speed | 13,400 rpm |
Max. RCF | 12,100 x g |
Display | Large LCD |
Height (English) Exterior | 5.12 in. |
Height (Metric) Exterior | 13.0 cm |
Noise Level | <49 dB(A) |
Timer | 15 sec. to 30 min |
Width (English) Exterior | 8.86 in. |
Width (Metric) Exterior | 22.5 cm |
Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
Includes | With Rotor F-45-12-11 |
Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids |
Ventilated | Yes |
Electrical Requirements | 230 V, 50/60 Hz (GB) |
Dimensions (L x W x H) Exterior | 226 x 239 x 130 mm |
Controller Type | Keypad |
Model | Centrifuge MiniSpin |
Wattage | 70 W |
Rotor Included | Yes |
Refrigerated | No |
Standards | EN ISO 12100, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 50581 (refer to manual for more details) |
Frequency | 50/60 Hz |
Voltage | 230 V |