Gibco™ Embryonic Stem Cell FBS, qualified, US origin
Fetal Bovine Serum, ES cell qualified, is specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.
Additional Details:
Additional Details: Weight: 0.05000kg
Disclaimers: For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Code. 15946942
Quantity Price
1 £ 72.5 / 50mL
In stock, ready to ship: 21
Specification
Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Tested For Bovine Respiratory Syncytial Virus FA, Cytopathic Agents, Hemadsorbing Agents, Reovirus FA, Rabies FA, Hemoglobin (ng/dL), Blue Tongue Virus, Bovine Adenovirus FA, Bovine Parvovirus FA, Osmolality, Sp2 Performance Assay, Electrophoretic Pattern, Total Protein, Endotoxin (EU/mL), Mycoplasma, Sterility Testing (Bacteria and Fungi)
Donor Herd No
Endotoxin Concentration <50 EU/mL
Hemoglobin Concentration ≤25 mg/dL
Purity or Quality Grade ES Cell-Qualified
Product Line One Shot™
Shipping Condition Frozen
Fetal Bovine Serum, ES cell qualified, is specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells. It is Gibco FBS quality and performance you trust offered in a single-use FBS aliquot, which provides greater convenience, saves time, and is easy to use.
  • Sera Category: Specialty
Alternative Products