Brannan™ Lab Thermometer
General purpose laboratory thermometer without calibration certificate.
Additional Details:
Additional Details: Weight: 0.20000kg
Product Code. 16019961
Quantity Price
1 £ 8.55 / Each
Estimated Shipment
21-08-2025
Specification
Immersion Type Partial Immersion
Length (Metric) 305 mm
Material Glass
Thermometer Fill Red Spirit
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 14°F
Min. Temperature (Metric) -10°C
Temperature Range (English) 14°F to 122°F
Temperature Range (Metric) -10°C to +50°C
Accuracy ±1.0°C at 35°C
Subdivision 1°C
Width (Metric) 1.2 cm
  • Lab thermometer
  • Safe non-mercury filling
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Alternative Products