622.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.
Autoclavable | Not Autoclavable |
---|---|
Capacity (Metric) | 1.5 mL |
Color | Natural |
Material | Polypropylene (Virgin) |
Includes | NonKnurl |
Packaging | 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag. |
Temperature Range (Metric) | Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage |
Clean Claims | Non-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor |
Binding Property | Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition |
Max. Torque | Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm |
Sterility | Non-sterile |
Graduated | Yes |
Closure Type | Screw Cap with O-ring (Unattached) |
Fit for Purpose | PCR Ready |
Bottom Shape | Non-skirted |
Max. RCF | 25,000 x g |
Product Type | Screw-cap Microtube |