432.57 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Use promo code "24095" to get your promotional price.
Certifications/Compliance | Declaration of Conformity |
---|---|
Autoclavable | Not Autoclavable |
Material | Metal Ejector |
Model | P8x10G |
Number of Channels | 8 |
Product Line | Pipetman G |
Type | Manual Air Displacement Pipette |
Volume (Metric) | 1 to 10 μL |
Warranty | 3 Years |
Length (English) | 5.98 in. |
Width (English) | 12.87 in. |
Height (English) | 1.54 in. |
Color Code | Red |