Epredia™ PrintMate™ Embedding Cassettes
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Additional Details:
Weight:
4.53600kg
Specification
Pore Style | Round Holes |
---|---|
Closure Included | No lids |
Quantity | 4 x 250 cassettes (1000)/cs |
Closure Type | Reversible |
PrintMate Compatible | Yes |
Color | Lilac |
For Use With (Equipment) | For Use with Metal Lids |
Product Type | Embedding Cassette |