3.01 GBP valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Use promo code "24097" to get your promotional price.
Product Type | Hooded Coveralls |
---|---|
Size | 2X-Large |
Material | Polyethylene |
Color | White |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Closure Type | Zipper |
Seam Type | Stitched |
Protection Class | Category III, Type 5 and 6 |
Certifications/Compliance | (EU) 2016/425, EN 1073-2, EN 1149-1 |
Size (Metric) Chest | 116 to 124 cm |