Memmert™ Interior Ip68 Socket (For Temperature Restriction +70°C Option) 230V, 2. A, For Icp110
Additional Details:
Weight:
0.01100kg
Specification
For Use With (Equipment) | Can be Switched Off with The On/Off Switch, Cannot be Switched Individually |
---|---|
Temperature Range (Metric) | 70°C |
Type | Interior Socket |
Voltage | 230 V |
Amperage | 2.2 A |