Eppendorf™ CellXpert™ C170i, Segmented Inner Door, Oxygen Control
CellXpert™ C170i CO2 incubator provides reliable growth conditions for more reproducible results.
Additional Details:
Weight:
190.50000kg
Specification
Capacity (Metric) | 170 L |
---|---|
Chamber Material | Stainless Steel |
CO2 Sensor Technology | IR Sensor |
Oxygen Control | Yes |
Plug Type | GB |
Temperature Range (Metric) | Ambient +4° to 50°C |
Display | VisioNize™ touch interface |
Door Style | Right-Sided Handle |
Includes | Inner door with 4 segments |
O2 Sensor Technology | ZrO2 (zirconium dioxide) |
Ports | 2 |
Dimensions (D x W x H) Interior | 44.5 x 53.9 x 69.2 cm (17.5 x 21.2 x 27.2 in.) |
Dimensions (L x W x H) | 71.5 x 71.8 x 90.0 cm (28.3 x 28.1 x 35.4 in.) |
Electrical Requirements | 220 to 240 V, 50/60 Hz |