850.01 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Connection Type | RS232 (one port included) |
---|---|
Description | User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator |
Capacity | 4200.00 g |
Display Type | Backlit LCD |
Readability | 0.0100 g |
For Use With (Application) | Basic Weighing, Parts Counting, Percent Weighing |
Linearity | 0.0200 g |
Model | PR4202/E |
Repeatability | 0.0100 g |
Stabilization Time | 1.000 s |
Weighing Modes | Milligram, Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Newton, & Grain |
Dimensions (L x W x H) | 55.000 x 39.000 x 29.000 cm |
Length (English) | 12.50 in. |
Length (Metric) | 317.00 mm |
Width (English) | 7.90 in. |
Width (Metric) | 201.00 mm |
Height (English) | 3.70 in. |
Height (Metric) | 93.000 mm |
Depth (English) | 12.50 in. |
Depth (Metric) | 317.00 mm |
Weight (English) | 7.700 lb. |
Weight (Metric) | 3.500 kg |
Electrical Requirements | AC Adapter (included) |