BRAND™ Multi-Channel Micropipette Transferpette™ S -8, adjustable, DE-M
Efficient: one-hand operation. Flexible: ergonomic design for all hand sizes. Reliable: volume-change protection prevents accidental adjustments.
Overview
502.81 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.54360kg
Product Code. 16437815
Quantity Price
1 £ 741.0 / Each
Estimated Shipment
30-05-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
Description and Specification
Specification
Certifications/Compliance DIN EN 285
Description Transferpette™ S-8, adjustable volume, DE-M, M8-50, 5-50 μL
Autoclavable Autoclavable
Material Polypropylene
Product Line Transferpette S
Sterility Non-sterile
Type Pipette
Volume (Metric) 5 to 50 μL
Temperature (Metric) Operating 15 to 40°C
Warranty 36 months
Length (Metric) 23.7 cm
Width (Metric) 14 cm
Height (Metric) 12.3 cm
The Transferpette™ S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette™ S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.
  • The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment.
  • The Transferpette™ S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools.
  • A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.