BINDER™ Overheating Alarm
For Use With: BF 56, BF 115, BF 260, BF 720
Additional Details:
Weight:
0.00100kg
Specification
Description | Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device |
---|---|
For Use With (Equipment) | BINDER Chambers BF 56, BF 115, BF 260, BF 720 |
Material | Plastic |
Type | Alarm |