BINDER™ Inert Gas Connection
For Use With: BF 56
Additional Details:
Weight:
0.00100kg
Specification
Description | Inert gas connection - With gas inlet and outlet, Ø 10 mm, incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option |
---|---|
For Use With (Equipment) | BINDER Chambers BF 56 |
Material | Steel |
Type | Gas Connection |
Diameter (English) | 0.4 in. |
Diameter (Metric) | 10 mm |