BINDER™ Left Access Port (10 to 30 mm)
For Use With: BF 56, BF 115, BF 260, BF 400, BF 720
Additional Details:
Weight:
0.28000kg
Specification
Description | Access port with silicone plug - 10 mm, left |
---|---|
For Use With (Equipment) | BINDER Chambers BF 56, BF 115, BF 260, BF 400, BF 720 |
Material | Steel |
Type | Access Port |
Includes | Silicone plug |
Diameter (Metric) Inside | 10 mm |