BINDER™ Temperature Analog Output
For Use With: BF 56, BF 115, BF 260, BF 720
Additional Details:
Weight:
0.00100kg
Specification
Description | Analog output 4-20 mA - for temperature values (output not adjustable) |
---|---|
For Use With (Equipment) | BINDER Chambers BF 56, BF 115, BF 260, BF 720 |
Material | Plastic |
Type | Analog Output |
For Use With (Application) | Temperature |