Use promo code "24112" to get your promotional price.
Invitrogen™ Platinum™ SuperFi II DNA Polymerase
Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR.
657.36 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Additional Details:
Weight:
0.32000kg
Specification
Content And Storage |
• Platinum SuperFi II DNA Polymerase (500 μL) • 5X SuperFi II Buffer (6 x 1.25 mL) Store at –20°C. |
---|---|
GC-Rich PCR Performance | High |
Polymerase | Platinum SuperFi II DNA Polymerase |
Reaction Speed | Fast |
Quantity | 500 reactions |
Shipping Condition | Dry Ice |
For Use With (Application) | Hot-start PCR,High-fidelity PCR |
For Use With (Application) | Hot-start PCR,High-fidelity PCR |
Fidelity (vs. Taq) | >300X |
Hot Start | Built-In Hot Start |
No. of Reactions | 500 Reactions |
Overhang | Blunt |
Reaction Format | Standalone |
Size (Final Product) | 20 kb or less |