70.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Material | Stainless Steel (304) |
Weight (Metric) | 0.040 kg |
Mass | 200 mg |
Tolerance | 0.020 mg |
Includes | Protective Case |