Sartorius Entris™ II Advanced Precision Balances
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
2189.75 GBP valid until 2025-12-31
Use promo code "24377" to get your promotional price.
Use promo code "24377" to get your promotional price.
Additional Details:
Weight:
4.60000kg
Specification
Certifications/Compliance | CE Certificate, CSA for Canada |
---|---|
Connection Type | D-sub connector, 9-pin; USB UTL; USB Device |
Description | Without Internal Calibration |
Capacity | 420 g |
Display Type | Intuitive Graphic Touch Technology |
Readability | 1 mg |
Material | Glass, PBT, Stainless Steel |
Model | Electronic Precision Balance |
Stabilization Time | ≤1.0 seconds |
Type | Balance |
Voltage | 100/240 V |
Amperage | 0.2 A |
Frequency | 50/60 Hz |
Legal for Trade | Class I |