Honeywell Fluka Sodium hypochlorite solution, Purum, ∼10% (RT), Honeywell Fluka™
Additional Details:
CAS Number:
7681-52-9
Weight:
30.75000kg
Transport:
UN number : 1791
Chem class : 8
Pack group : II
Specification
Boiling Point | 111°C |
---|---|
Chemical Name or Material | Sodium hypochlorite solution |
Melting Point | -30°C to -20°C |
CAS | 7732-18-5 |
Color | Clear |
CAS | 7681-52-9 |
Physical Form | Liquid |
Quantity | 25 L |
Molecular Formula | NaClO |
UN Number | UN1791 |
Molecular Weight (g/mol) | 74.44 g/mol |
Grade | Synthesis |