Labcon SuperClear™ 17 x 100 mm Culture Tubes
For cell culture and general laboratory use
Additional Details:
Weight:
6.80390kg
Specification
Closure Material | Polyethylene |
---|---|
Diameter (Metric) | 17 mm |
Max. RCF | 1500 RCF |
Product Type | Round-bottom Tube |
Autoclavable | Not Autoclavable |
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity |
Closure Type | Dual-position Cap |
Color | Clear |
Length (Metric) | 100 mm |
Material | Polystyrene |
Disposable | Single-use |
Graduated | No |
For Use With (Application) | Culture |
Length (English) | 3.93 in. |
Packaging Quantity | 125 Tubes/Bag, 8 Bags/Cs. |
Quantity | 125 Tubes/Bag |
For Use With (Equipment) | For Cell Culture and General Laboratory Use |
Product Line | SuperClear™ |
Sterility | Sterile |
Capacity (Metric) | 14 mL |
Bulk Pack | 125 |