A&D Weighing™ Apollo Analytical Balance with External Calibration
Apollo Analytical Series improve productivity and efficiency of your workflows with user-friendly and innovative features. It is designed to provide reliable results across a range of environments and applications.
4023.25 GBP valid until 2025-12-31
Use promo code "24305" to get your promotional price.
Use promo code "24305" to get your promotional price.
Additional Details:
Weight:
7.00000kg
Specification
Certifications/Compliance | GLP/GMP/GCP/ISO |
---|---|
Connection Type | RS-232 C and USB |
Capacity | 320 g |
Display Type | Reverse-backlit LCD |
Readability | 0.0001 g |
Material | Stainless Steel |
Model | GF-324A |
Stabilization Time | 2 seconds (300 g)/1.5 seconds (100 g) |
Electrical Requirements | 30 VA to adapter (12 V, 1 A) |