Eppendorf™ Centrifuge 5910 Ri
Centrifuge 5910 Ri stands out with an intuitive touch screen interface and a range of innovative solutions for usability, documentation and user management. Its wide variety of fixed-angle and swing-bucket rotors accommodate a comprehensive spectrum of applications in a compact footprint.
11570.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
133.00000kg
Specification
Max. RCF | 22,132 x g |
---|---|
Refrigerated | Yes |
Display | VisioNize™ touch interface |
Includes | Rotor |
Model | Centrifuge 5910 Ri |
Noise Level | <59 dB(A) with Rotor FA-6 x 50 |
Rotor | FA-6x50 |
Rotor Included | Yes |
Timer | 10 s to 99 h 59 min, with continuous run function, short-spin |
Type | Benchtop Centrifuge |
Width (English) Exterior | 28.3 in. |
Width (Metric) Exterior | 72 cm |
Height (English) Exterior | 14.5 in. |
Height (Metric) Exterior | 37 cm |
Depth (English) | 25.9 in. |
Depth (Metric) | 68 cm |
Electrical Requirements | 230 V/50-60 Hz (GB) |
Voltage | 230 V |
Frequency | 50/60 Hz |