Agilent BioTek MultiFlo FX Multi-Mode Dispenser
MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates.
Additional Details:
Weight:
16.00000kg
Specification
Accuracy | ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL |
---|---|
Product Type | Microplate Dispenser |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Volume (Metric) | 500 nL to 30,000 μL |
For Use With (Application) | Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization |
For Use With (Equipment) | BioStack and third party automation compatible; BioSpa 8 Automated Incubator compatible |
Includes | RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing |
Interface | Color touch screen; Create, edit or run multiple protocols onboard |
Model | MFXP2R |
Plate Type | Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) |
Precision | <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL |
Software | LHC2 Software; LHC2 Secure for 21 CFR Part 11 compliance (option); SiLA Compliant driver (option) |
Speed | 96 wells, 5 μL cass, 10 μL/ well: 3 s, 96 wells, 5 μL cass, 20 μL/ well: 3.5 s, 384 wells, 5 μL cass, 5 μL/ well: 6.5 s, 384 wells, 10 μL cass, 10 μL/ well: 8 s, 384 wells, 1 μL cass, 1 μL/ well: 6 s, 1536 wells, 1 μL cass, 1 μL/ well: 21 s |
Width (English) | 17.2 in. |
Width (Metric) | 43.5 cm |
Height (English) | 8 in. |
Height (Metric) | 20.3 cm |
Depth (English) | 11.8 in. |
Depth (Metric) | 29.2 cm |
Weight (English) | 14.3 lb. |
Weight (Metric) | 6.5 kg |
Electrical Requirements | 90 W max. |
Voltage | 100/240 V |