Labcon™ PerformR™ 15 mL Polystyrene Centrifuge Tubes
Made of crystal clear resin and featuring bold, etched graduations, these tubes can be used for applications that require a glass clear material.
Additional Details:
Weight:
1.07720kg
Specification
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
---|---|
Closure Material | Polyethylene, LDPE Foam |
Closure Color | Purple |
Color | Natural |
Capacity (Metric) | 15 mL |
Disposable | Single-use |
Material | Polystyrene |
For Use With | Centrifuges, Rotators, Shakers |
Packaging | 25 Tubes/Rack, 2 Racks/Pk., 10 Pk./Cs. |
Quantity | 125 Tubes/Bag, 4 Bags /Cs. |
Sterility | Sterile |
RNase Free | Yes |
Dnase Free | Yes |
Autoclavable | Not Autoclavable |
Shape | Cylindrical |
Length (Metric) | 118 mm |
Clean Claims | ATP, RNase/DNase and Endotoxin-free |
Bottom Shape | Conical |
Closure Type | Flat Screw Cap |
Closure Included | Yes |
Writing Area | Yes |
Diameter (Metric) | 17 mm |
Graduated | Yes |
Graduations | Molded |
Product Type | Centrifuge Tube |
Max. RCF | 3,000 x g |